Learn More
AdipoGen Fridamycin A
SDP
Supplier:  AdipoGen BVT0469M005
Encompass_Preferred
Chemical. CAS 30270-05-4. Formula C25H26O10. MW 486.5. Isolated from Streptomyces parvulus. C-glycosidic angucyclinone derivative. Antibiotic. Antitumor agent. Antibacterial agent. Isomer of fridamycin B Prod. No. BVT-0470.
Catalog No. 50-256-1889