Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, 38mm OD Delong Top, WITHOUT Baffles, Approx OD x Height (mm): 210 x 260
SDP
Supplier:  Chemglass Life Sciences CLS202011
Encompass_Preferred
Flask, Fernbach, 2800mL, 38mm OD Delong Top, WITHOUT Baffles, Approx OD x Height (mm): 210 x 260. Flasks are designed for culturing organisms that require a large surface area to volume ratio.
Catalog No. 50-121-4631