Learn More
Perkin Elmer US LLC Front Surface Accessory
Supplier:  Perkin Elmer US LLC 52123130
Encompass_Preferred
With the simple-to-use Front Surface Accessory, fluorescence and phophorescence measurements can be taken by placing samples directly on the accessory. You can, however, use the included powder window sample holders (52123164) for solid samples or cuvettes (not included) to measure small sample sizes (under 10 mm) of opaque or turbid samples.
Catalog No. 50-211-3289
Only null left
Add to Cart
Request a Quote