Learn More
Medchemexpress LLC HY-B1658A 1mg Medchemexpress, Frovatriptan (succinate hydrate) CAS:158930-17-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1658A1MG
Encompass_Preferred
Medchemexpress, HY-B1658A 1mg Frovatriptan (succinate hydrate) CAS:158930-17-7 Frovatriptan succinate hydrate is a potent, high affinity, selective and orally active 5-HT1B, HT1D receptor agonist and a moderately potent 5-HT7 receptor agonist, with pKi values of 8.6, 8.4, and 6.7, respectively. Frovatriptan succinate hydrate is effective in treating the full spectrum of migraine including the associated symptoms of nausea, vomiting, photophobia, and phonophobia. Frovatriptan succinate hydrate can also be used as in mini-prophylaxis in menstrual migraine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0155