Learn More
Medchemexpress LLC Frovatriptan succinate hydrate | 158930-17-7 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1658A5MG
Encompass_Preferred
Frovatriptan succinate hydrate ((R)-Frovatriptan succinate hydrate) is a potent, high-affinity, selective, and orally active agonist for 5-HT1B (pK50 of 8.2) and 5-HT1D receptors. It demonstrates over 10-fold selectivity for 5-HT1B and 5-HT1D receptors compared to 5-HT1A, 5-HT1F, and 5-HT7, and more than 1000-fold selectivity over other 5-HT, dopamine, histamine H1, and α1-adrenoceptors. This compound has potential for use in migraine research.
  • Potent, high-affinity, selective 5-HT1B and 5-HT1D receptor agonist.
  • Orally active.
  • Exhibits >10-fold selectivity for 5-HT1B and 5-HT1D over 5-HT1A, 5-HT1F, and 5-HT7.
  • Potential for migraine research.
  • Oral bioavailability of 22%-30%, not affected by food. Long terminal half-life of approximately 26 hours. Metabolized primarily by CYP1A2.
Catalog No. 50-004-13170