Learn More
Cell Biologics Rat Primary Dermal Fibroblasts, Frozen Vial
SDP
Supplier:  Cell Biologics RN6068FROZEN
Encompass_Preferred
Rat Primary Dermal Fibroblasts can be used in assays of standard biochemical procedures performed with cell cultures include RT-PCR, Western blotting, immunoprecipitation, immunofluorescent flow cytometry, or generating cell derivatives for desired research applications.
Catalog No. NC1857706