Learn More
Biochain Institute Inc Multiple Species Frozen Tissue Array - Liver, 1 slide/PK
SDP
Supplier:  Biochain Institute Inc T61341491
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. Multiple Species Frozen Tissue Array is designed for testing the species reactivity of antibodies. This array can bring very important information of antibodies in immunohistochemistry studies. Besides, frozen tissues are always the solution in case antibodies can not recognize their antigens in paraffin embedded tissues. The Multiple Species frozen tissue array can also enable you to study expression of genes or proteins of interest among different animals. BioChain's Frozen Tissue Arrays are manufactured using high-quality tissues. Each section is approximately 5 um thick. H.E. staining and immunostaining using beta-actin antibody ensure quality. Frozen tissue array is ideal for rapid cellular localization of RNA and protein expression.
Catalog No. 50-180-889