Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Mouse Whole Brain Segmentation Panel, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T6334035
Encompass_Preferred
Whole brain segmentation panels are designed for study whole brain of small animals. The animals' whole brain was procured intact and was sliced into 6 equal coronal regions. Each region is 2 mm thick. The 6 regions were embedded in the same OCT block. The frozen section panel cut from this block can enable scientists or research investigators do biochemical analysis, such as the determination of neurotransmitter and metabolite concentrations for reproducible brain regions (i.e. discrete terminal fields or nuclei).
Catalog No. 50-181-334