Learn More
MSC Cooper Bussmann FRS-R-350 300 VDC, 600 VAC, 350 Amp, Time Delay General Purpose Fuse Bolt-on Mount, 11-5/8" Overall Length, 20 at DC, 200 (RMS) kA Rating, 2-9/16" Diam
Supplier:  MSC 75908806
Encompass
Cooper Bussmann FRS-R-350 300 VDC, 600 VAC, 350 Amp, Time Delay General Purpose Fuse Bolt-on Mount, 11-5/8" Overall Length, 20 at DC, 200 (RMS) kA Rating, 2-9/16" Diam
Catalog No. 50-130-5056