Learn More
MSC Cooper Bussmann FRS-R-6 300 VDC, 600 VAC, 6 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam
Supplier:  MSC 75908921
Encompass
Cooper Bussmann FRS-R-6 300 VDC, 600 VAC, 6 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam
Catalog No. 50-130-5065