Learn More
ABclonal Technology FRS2 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A23135PM20UL
Encompass_Preferred
Enables fibroblast growth factor receptor binding activity and neurotrophin TRKA receptor binding activity Involved in negative regulation of cardiac muscle cell differentiation Acts upstream of or within fibroblast growth factor receptor signaling pathway Located in adherens junction Biomarker of renal cell carcinoma
Catalog No. 50-000-06373