Learn More
Medchemexpress LLC Fructo-oligosaccharide DP14 | 137405-38-0 | 1 MG

Supplier: Medchemexpress LLC HYN80591MG
Fructo-oligosaccharide DP14 is an oligosaccharide that can be isolated from Atractylodes lancea. It is a natural product intended for research use only.
- Purity: 98.58%
- Molecular weight: 2287.98
- Formula: C84H142O71
- CAS no.: 137405-38-0
- Appearance: Solid
- Color: White to off-white
- Structure classification: Saccharides, Polysaccharides
- Initial source: Plants (Leguminosae, Erythrina sigmoidea Hua; Compositae)
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: 4°C, protected from light. In solvent: -80°C for 6 months; -20°C for 1 month (protected from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.