Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructo-oligosaccharide DP8/GF7 | 62512-21-4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN683610MG

Encompass_Preferred

Fructo-oligosaccharide DP8/GF7 is a type of fructooligosaccharide (FOS) with a degree of polymerization (DP) of 8. It consists of 7 fructose units linked by (2→1)-β-glycosidic bonds, featuring a single D-glucosyl unit. This product has a purity of 99.76%, a molecular weight of 1315.14, and the chemical formula C48H82O41. It appears as a white to off-white solid and is sourced from Gramineae Saccharum officinarum plants.

  • Degree of polymerization (DP) of 8
  • Comprised of 7 fructose units
  • Single D-glucosyl unit at non-reducing end
  • Purity of 99.76%
  • Molecular weight 1315.14; formula C48H82O41
  • White to off-white solid appearance
  • Derived from Gramineae Saccharum officinarum
  • Soluble in H2O at 150 mg/mL
  • Store at 4°C, sealed, away from moisture and light

Catalog No. 50-004-20983


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.