Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Randox Fructosamine Control Level 1, Lyophilised, Human-based serum, 3 x 1 ml, Assayed, Control Level 1
SDP

Supplier:  Randox FR2994

Encompass_Preferred

The Randox Acusera Fructosamine Control Level 1 (FR2994) is designed for in vitro diagnostic use, specifically to monitor the accuracy and precision of fructosamine assays on clinical chemistry systems. It features enhanced stability and minimizes waste, with a working stability lasting to its expiry at 2°C to 8°C, and a reconstituted stability of 28 days under the same conditions. It is ideal for facilities that require reliable quality control for diabetes monitoring.

  • Engineered for in vitro diagnostic use
  • Monitors accuracy of fructosamine assays
  • Enhanced stability to expiry at 2°C to 8°C
  • 28-day reconstituted stability at 2°C to 8°C
  • Designed for quality control in clinical settings
  • Matrix: Human based serum
  • Volume: 3 x 1 ml
  • Assayed values for reliable reporting
  • Suitable for diabetes monitoring applications

Catalog No. NC0766326


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.