Learn More
U.S. Pharmacopeia Fructose, 57-48-7, MFCD00148910, 125mg
Supplier:  U.S. Pharmacopeia 1286504
Encompass
Molecular formula C6H12O6, Molecular weight 180.16, Melting Point 217.4 - 221 °F (103 - 105 °C) (decomposes), Synonyms: Fruit sugar, D-Fructose
Catalog No. NC9352893