Learn More
Medchemexpress LLC Fructose-alanine (mixture of diastereomers) | 16124-24-6 | MFCD31562994 | >95.0% | 251.23 g/mol | C9H17NO7 | 1 MG

Supplier: Medchemexpress LLC HYW5859331MG
Fructose-alanine (mixture of diastereomers) is an Amadori compound used as an aroma precursor and research chemical in flavor, fragrance, and food chemistry studies. It is supplied as a small-quantity reference material for analytical and biochemical work.
- Amadori compound used as an aroma precursor in flavor and fragrance research.
- Provided as a small-quantity research standard suitable for analytical studies.
- CAS number 16124-24-6 for unambiguous identification.
- Chemical formula C9H17NO7 and molecular weight 251.23 g/mol.
- Purity greater than 95.0% (HPLC) when specified by suppliers.
- Typically supplied as a pale yellow solid; follow supplier storage and handling instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.