Learn More
Medchemexpress LLC Fructose (standard) | 7660-25-5 | MFCD00065409 | 180.16 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0395R50MG
Encompass_Preferred
Fructose (Standard) is an analytical standard of Fructose, intended for research and analytical applications. It is a simple ketonic monosaccharide found in many plants, often bonded to glucose to form sucrose. This compound is a reference standard supplied assay.
  • Used as an analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-90027