Learn More
Cayman Chemical Fructose1 6bIsphosphatse1 50mg
SDP
Supplier:  Cayman Chemical 1886050 MG
Encompass_Preferred
A cell-permeable benzoxazolo-sulfonamide compound that blocks human FBPase-1 enzymatic activity by competing at the AMP allosteric binding site (IC50 = 3.4 µM; Ki = 1.1 µM); blocks glucose production in starved rat hepatoma cells (IC50 = 6.6 µM)
Catalog No. 50-298-4407