Learn More
Medchemexpress LLC Fructosyl-lysine | 21291-40-7 | 99.9% | 25 MG

Supplier: Medchemexpress LLC 25MGHY129380
Fructosyl-lysine is an Amadori glycation product formed from glucose and lysine via the Maillard reaction. It serves as a precursor to glucosepane, a lysine-arginine protein cross-link that can indicate diabetes. This product is for research use only.
- Acts as a precursor to glucosepane
- Can be phosphorylated and converted to glucose 6-phosphate
- Supports E. coli growth
- Increased levels observed in diabetic rats
- Molecular Weight: 308.33
- Formula: C12H24N2O7
- Appearance: Light yellow to brown solid
- Solubility in H2O: 50 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.