Learn More
Medchemexpress LLC Fruquintinib-d6 | 2978508-76-6 | 99.5% | 399.43 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY19912S5MG
Encompass_Preferred
Fruquintinib-d6 is the deuterium labeled Fruquintinib (HY-19912). Fruquintinib (HMPL-013) is a highly potent and selective VEGFR 1/2/3 inhibitor with IC50s of 33, 0.35, and 35 nM, respectively. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-01805