Learn More
Selleck Chemical LLC Fruquintinib (HMPL-013) 5mg 1194506-26-7
SDP
Supplier:  Selleck Chemical LLC S56675MG
Encompass_Preferred
Fruquintinib (HMPL-013) is a small molecule inhibitor with strong potency and high selectivity against VEGFR family. It inhibits VEGFR 1, 2, 3, with IC50 values of 33 nM, 35 nM and 0.5 nM, respectively and shows only weak inhibition of RET, FGFR-1 and c-kit kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8251