Learn More
Fry’s Reagent, Ethanolic Etchant for Austenite Stainless Steel (Meets ASTM E407 Specification Etchant 79), Reagents
Fry’s Reagent, Ethanolic Etchant for Austenite Stainless Steel (Meets ASTM E407 Specification Etchant 79), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$136.00 - $318.00
Chemical Identifiers
| CAS | 7732-18-5 , 7647-01-0 , 64-17-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-170
|
Reagents Holdings Llc
CF1800001C |
1 L |
Each for $318.00
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
01-335-169
|
Reagents Holdings Llc
CF180000500C |
500 mL |
Each for $136.00
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.