Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fry’s Reagent, Ethanolic Etchant for Austenite Stainless Steel (Meets ASTM E407 Specification Etchant 79), Reagents

Catalog No. 01335170
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-170 1 L
01-335-169 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-170 Supplier Reagents Holdings Llc Supplier No. CF1800001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fry’s Reagent, Ethanolic Etchant for Austenite Stainless Steel (Meets ASTM E407 Specification Etchant 79), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 64-17-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Green
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0784
pH <1
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Fry’s Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.