Learn More
Andwin Scientific FRZR BOX DIVIDER 100 HOLE 12PK
SDP
Supplier:  Andwin Scientific 6161590
Encompass_Preferred
The Freezer Box Divider 100 Hole is used to create partitions in the cardboard freezer box. This product can be used to hold up to 100 cryogenic vials stored in liquid nitrogen or mechanical freezers. It is sold in a 12-pack.
Catalog No. 50-727-641