Learn More
Andwin Scientific FRZR BOX DIVIDER 64 HOLE 12/PK
SDP
Supplier:  Andwin Scientific 6161575
Encompass_Preferred
The Freezer Box Divider 64 Hole is used to create partitions in the cardboard freezer box. This product can be used to hold up to 64 cryogenic vials stored in liquid nitrogen or mechanical freezers. It is sold in a 12-pack.
Catalog No. 50-727-638