Learn More
Andwin Scientific FRZR BOX DIVIDER 81 HOLE 12/PK
SDP
Supplier:  Andwin Scientific 6161580
Encompass_Preferred
The Freezer Box Divider 81 Hole is used to create partitions in the cardboard freezer box. This product can be used to hold up to 81 cryogenic vials stored in liquid nitrogen or mechanical freezers. It is sold in a 12-pack.
Catalog No. 50-727-639