Learn More
Chemglass Life Sciences Freezer Rack Only, Upright Drawer Style, for Standard 2" Boxes, Box Capacity: 3L x 3H, 16-1/2"L x 5-1/2"W x 7-1/8"H
SDP
Supplier:  Chemglass Life Sciences CLS3795001
Encompass
Freezer Rack Only, Upright Drawer Style, for Standard 2" Boxes, Box Capacity: 3L x 3H, 16-1/2"L x 5-1/2"W x 7-1/8"H. Drawers make these freezer racks easy and convenient to use. No need to remove the entire rack from the freezer to gain access, just slide the drawer out. Centrifuge tube racks are specifically designed to provide an alternative for storing large quantities of samples in 15mL and 50mL centrifuge tubes. Racks for two-inch boxes hold between nine and twenty-four boxes. High quality racks are constructed of corrosion-resistant stainless steel and include handles and identification card holders.
Catalog No. NC1510168