Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for Standard 2" Boxes, 7 box capacity
Supplier:  Cole-Parmer 0439844
Encompass_Preferred
These upright freezer racks are great for storing standard 2-inch cardboard, plastic or metal boxes. Designed to fit all major brands of f 80C ultralow upright freezers. The racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0494