Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for 96/384-Well Microtiter Plates; accepts 35 with lids or 42 without lids
Supplier:  Cole-Parmer 0440110
Encompass_Preferred
These racks come with locking rods to securely store microtiter plates in place. The racks have sturdy dual handles for easy handling, and made of corrosion-resistant stainless steel.
Catalog No. 50-196-0746