Learn More
Aviva Systems Biology Corp FSBP Antibody - N-terminal region (ARP32780_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32780_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FSBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FSBP. Alternate names: ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54760