Learn More
Medchemexpress LLC FSCPX 10mg | 156547-56-7 | 506.55 g/mol | C23H27FN4O6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11604210MG
Encompass_Preferred
FSCPX is a research-grade irreversible A1 adenosine receptor antagonist supplied for pharmacology studies. It is characterized by a molecular weight of 506.55 g/mol, molecular formula C23H27FN4O6S, and high reported purity; recommended storage and formulation guidance are provided for both powder and solution forms.
Catalog No. 50-002-27900