Learn More
Aviva Systems Biology Corp FSD1L Antibody - C-terminal region (ARP60577_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60577_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FSD1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FSD1L. Alternate names: CCDC10, CSDUFD1, FSD1CL, FSD1NL, MGC45564, MIR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-089