Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FSH beta protein, mouse (HEK293, Fc) | 95.0% | 42-60 kDa | 10 UG
SDP

Supplier:  Medchemexpress LLC HYP7794010UG

Encompass_Preferred

The FSH beta protein, in conjunction with the alpha chain CGA, forms follitropin, also known as follicle-stimulating hormone (FSH). This hormone is crucial for follicle development and spermatogenesis within reproductive organs. It binds to FSHR, a G protein-coupled receptor on target cells, initiating downstream signaling pathways. The intricate assembly of FSH beta within the hormone heterodimer underscores its essential contribution to reproductive processes and hormonal signaling cascades.

  • Appearance: Lyophilized powder
  • Formulation: Lyophilized from a 0.22 μm filtered solution of PBS, pH 7.4, 8% trehalose
  • Endotoxin level: <1 EU/μg, determined by LAL method
  • Storage: Stored at -20°C for 2 years from date of receipt
  • Stability after reconstitution: Stable at 4°C for 1 week or -20°C for longer (with carrier protein)

Catalog No. 50-003-99886


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.