Learn More
Selleck Chemical LLC FSLLRY-NH2 TFA S9915-25MG
SDP
Supplier:  Selleck Chemical LLC S991525MG
Encompass_Preferred
FSLLRY-NH2 TFA (FSY-NH2) is a selective protease-activated receptor 2 (PAR2) inhibitor with an ic50 of 50 M in PAR2-KNRK cells
Catalog No. 50-000-24709