Learn More
Medkoo Biosciences Inc FT-671 10MG
SDP
Supplier:  Medkoo Biosciences Inc 471006
Encompass
FT671 is a potent and selective USP7 inhibitor with high affinity and specificity in vitro and within human cells. CAS Number 1959551-26-8. MEDKOO BIOSCIENCES INC mfg# 471006
Catalog No. NC1894162