Learn More
Medchemexpress LLC FT709 | 2413991-74-7 | 99.8% | C23H22N4O7S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14596710MM/1ML
Encompass_Preferred
FT709 is a potent and selective inhibitor of USP9X, exhibiting an IC50 of 82 nM. USP9X has been associated with various cellular functions, including centrosome function, chromosome alignment during mitosis, EGF receptor degradation, chemo-sensitization, and circadian rhythms. This product is for research use only.
  • Potent and selective USP9X inhibitor
  • Exhibits an IC50 of 82 nM
  • Associated with centrosome function
  • Associated with chromosome alignment during mitosis
  • Associated with EGF receptor degradation
  • Associated with chemo-sensitization
  • Associated with circadian rhythms
  • Decreases levels of ZNF598 (ubiquitin E3 ligase) and centrosomal protein CEP55 in HCT116 cells
  • Suitable for research applications
Catalog No. 50-003-53312