Learn More
Medchemexpress LLC Ft709 | 2413991-74-7 | 99.8% | C23H22N4O7S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14596710MG
Encompass_Preferred
FT709 is a potent, selective small-molecule inhibitor of the deubiquitinase USP9X (IC50 ≈ 82 nM), intended for biochemical and cell-based research to probe ubiquitin-dependent pathways and mitotic regulation. Supplied as a dry solid and as DMSO solution formats for flexible use in laboratory assays.
  • Potent, selective inhibition of USP9X (IC50 ≈ 82 nM).
  • Suitable for biochemical and cell-based assays.
  • Available as dry solid and DMSO solution for flexible dosing.
  • High reported purity for research applications.
  • Useful for studying mitosis and ubiquitin-dependent signaling.
Catalog No. 50-002-59595