Learn More
Medchemexpress LLC FT709 | 2413991-74-7 | C23H22N4O7S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1459671MG
Encompass_Preferred
FT709 is a potent and selective USP9X inhibitor with an IC50 of 82 nM. USP9X has been linked to various cellular functions including centrosome function, chromosome alignment during mitosis, EGF receptor degradation, chemo-sensitization, and circadian rhythms. This product is for research use only.
  • Potent and selective USP9X inhibitor
  • IC50 of 82 nM
  • Linked to centrosome function
  • Linked to chromosome alignment during mitosis
  • Linked to EGF receptor degradation
  • Linked to chemo-sensitization
  • Linked to circadian rhythms
  • Decreases levels of ZNF598 (ubiquitin E3 ligase)
  • Decreases levels of centrosomal protein CEP55 in HCT116 cells
Catalog No. 50-003-53973