Learn More
ATCC 20 STR Profiling Authentication Service
Supplier:  ATCC 135XV20
Encompass
One complete human cell line authentication service, utilizing short tandem repeat (STR) profiling. Trained experts amplify 17 STR loci plus Amelogenin using Promega’s PowerPlex 18D System. A comprehensive analysis report interprets both karyotypically normal and abnormal cell lines, includes a electropherograms supporting the allele calls at each locus, known reference profiling against the ATCC STR database and a comprehensive interpretation of results. This order contains twenty sample kits, mailed upon purchase. Sample kit mailed upon purchase. Download the instructions for sample preparation and submission.
Catalog No. NC1339593