Learn More
ATCC 10 STR Profiling Authentication Services
Supplier:  ATCC 135XV10
Encompass
One complete human cell line authentication service, utilizing Short Tanden Repeat (STR) profiling. Trained experts amplify 17 STR loci plus Amelogenin using Promega’s PowerPlex 18D System. A comprehensive analysis report interprets both karyotypically normal and abnormal cell lines, includes a electropherograms supporting the allele calls at each locus, known reference profiling against the ATCC STR database and a comprehensive interpretation of results. This order contains ten sample kits, mailed upon purchase. Sample kit mailed upon purchase. Download the instructions for sample preparation and submission.
Catalog No. NC1552616