Learn More
Cayman Chemical FTase InhIbItr IItrIfluoro 5mg
SDP
Supplier:  Cayman Chemical 166075 MG
Encompass_Preferred
A cell-permeable analog of FPP that potently inhibits FTase (IC50 = 50-75 nM), preventing farnesylation of Ras; does not inhibit geranylgeranyl transferase at similar concentrations (IC50 > 100 µM); blocks Ras-mediated transformation of NIH 3T3 cells
Catalog No. 50-296-6772