Learn More
Medchemexpress LLC HY-101787 50mg Medchemexpress, FTBMT CAS:1358575-02-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10178750MG
Encompass_Preferred
Medchemexpress, HY-101787 50mg FTBMT CAS:1358575-02-6 FTBMT is a selective GPR52 agonist with an EC 50 of 75 nM [1] . Antipsychotic and procognitive properties [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2216