Learn More
Medchemexpress LLC FTI-2153 (TFA) | 2820151-01-5 | 99.1% | 580.62 g/mol | C27H31F3N4O5S | 25MG
SDP
Supplier:  Medchemexpress LLC HY123242A25MG
Encompass_Preferred
FTI-2153 TFA is the trifluoroacetic acid salt of a potent and selective farnesyltransferase inhibitor developed for biochemical and cellular research. It demonstrates nanomolar potency (FTase IC50 = 1.4 nM), high reported purity, and is supplied as a solid or in solution for research applications. Storage recommendations and handling guidance are provided in the product safety data.
Catalog No. 50-002-35299