Learn More
Perkin Elmer US LLC FT-IR In-Service Lubricants Application Pack
Supplier:  Perkin Elmer US LLC L1608009
Encompass_Preferred
The FT-IR In-Service Lubricants Application Pack contains all of the resources and components necessary to perform efficient lubricant analysis in the laboratory and out in the field. The In-Service Lubricants Application Pack contains: Flow cell with ZnSe windows, a wedged spacer, and Leur fittings 3 mL Leur Lok plastic syringe starter pack of 100 IR Liquid Cell Leur connected waste tube Teflon plugs (pack of 6) In-Service Lubricants Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support the condition monitoring of in-service lubricants and provide a simplified workflow-driven operating environment; for faster, more reproducible analyses. The Touch Apps software supports the following analyses: JOAP direct-trending methods for petroleum, synthetic lubricants, and hydraulic fluids. ASTM E2412 direct-trending and spectral-subtraction methods. PerkinElmer methods for mineral and synthetic oils, petroleum hydraulic fluids, and turbine fluids. With
Catalog No. 50-211-7239
Add to Cart
Request a Quote