Learn More
Cole-Parmer Fitting, 316L Stainless Steel, Tee, Sanitary Clamp Union, 3/4
Supplier:  Cole-Parmer 5011835
Encompass_Preferred
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.
Catalog No. 50-255-7753