Learn More
Medchemexpress LLC FTO-IN-7 | 2243736-37-8 | 99.2% | 376.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147011100MG
Encompass_Preferred
FTO-IN-7 is an inhibitor of FTO (fat mass and obesity-associated protein) with an IC50 of <1 μM. It can be used for the study of small-cell lung cancers and human bone marrow striated muscle cancer. This product is for research use only.
  • Inhibitor of FTO (fat mass and obesity-associated protein)
  • Used for small-cell lung cancers and human bone marrow striated muscle cancer studies
  • Solid appearance
  • White to off-white color
Catalog No. 50-003-60425