Learn More
Medchemexpress LLC Fto-in-7 (compound 17) | 2243736-37-8 | MFCD34593645 | 99.2% | 376.24 g/mol | C18H15Cl2N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1470115MG
Encompass_Preferred
FTO-IN-7 is a small-molecule inhibitor of FTO (fat mass and obesity-associated protein) developed for research use. It has sub-micromolar inhibitory activity and is used in preclinical studies investigating cancer models, including small-cell lung cancer and human bone marrow striated muscle cancer.
  • Selective FTO inhibition with sub-micromolar potency.
  • High purity (99.22%) suitable for biochemical assays.
  • Good solubility in DMSO (200 mg/mL) for solution preparation.
  • Available in multiple milligram pack sizes for screening and dose-response work.
  • Stable when stored cold and protected from light.
Catalog No. 50-002-22622