Learn More
Medchemexpress LLC FTO-IN-8 | 2640366-38-5 | C19H23ClN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15110650MG
Encompass_Preferred
FTO-IN-8 is an N6-methyladenosine demethylase (FTO) inhibitor with an IC50 value of 5.5 μM. This compound exhibits anti-cancer cell proliferative activity, making it suitable for research into cancer targeted therapy and metabolism.
  • Inhibits proliferation of cancer cells (AGS, SNU-16, KATOIII)
  • Shows no cytotoxicity to normal colonic cells (CCD-841CoN)
  • Increases m6A and m6Am levels
  • Inhibits Wnt/PI3K-Akt signaling in gastric cancer AGS cells
Catalog No. 50-003-75777