Learn More
Cayman Chemical FTOhumn Monclonal ntIbo 100ug
SDP
Supplier:  Cayman Chemical 29020100 UG
Encompass_Preferred
Immunogen: Full-length recombinant human FTO protein • Host: Mouse • Species Reactivity: (+) Human • Applications: ELISA, IF, WB • MW = 58 kDa
Catalog No. 50-294-0582