Learn More
Medchemexpress LLC FTSDVSKQMEEEAVRLFIEWLKNGGPSSGAPPPS | 98.01% | 3692.15 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP12295MG
Encompass_Preferred
FTSDVSKQMEEEAVRLFIEWLKNGGPSSGAPPPS is an Exendin-4 peptide derivative that functions as a pure GLP-1 receptor agonist. These derivatives are structurally related to Exendin-4 and may also act as dual GLP-1/glucagon receptor agonists. They are of interest for medical use, particularly in the treatment of metabolic syndrome disorders such as diabetes and obesity, and for reducing excessive food intake.
  • Acts as a pure GLP-1 receptor agonist
  • Structurally derived from Exendin-4
  • Potential as a dual GLP-1/glucagon receptor agonist
  • May be used to treat metabolic syndrome disorders, including diabetes and obesity
  • Can help reduce excess food intake
  • Shows reduced activity on the GIP receptor to mitigate hypoglycemia risk
Catalog No. 50-004-22154